Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water for molecular biology

Supplier: Thermo Scientific Chemicals
Synonyms: Dihydrogen oxide

32739-0010 32739-0050
TS32739-0050EA 272 USD InStock
TS32739-0050 TS32739-0010
Water for molecular biology
Water


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now
Specification Test Results
Appearance Clear liquid
Additional info residue (at bottling): < 0.0001%
Additional info resistivity (at time of manufacture): > 18 megaohm
Additional info TOC: < 50 ppb
Color scale ≤5 APHA
Dnase,Rnase,Protease activity none detected
Chloride (Cl) ≤0.1 ppm
Sulfate (SO4) ≤0.5 ppm
Lead (Pb) ≤1 ppb

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty