Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Hydrogen peroxide 50% (w/w) in water stabilized, pure

Supplier: Thermo Scientific Chemicals
Danger

30286-0025 AC30286-5000
TS30286-5000EA 84 USD InStock
TS30286-5000 TS30286-0025
Hydrogen peroxide 50% (w/w) in water stabilized, pure
Hydrogen peroxide


Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 114 °C (760 mmHg)
Melting Pt: –52 °C
Density: 1.2 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II
Order Now
Specification Test Results
Appearance Clear colorless Solution
Refractive index 1.3665 to 1.3685 (20°C, 589 nm)
Density (20°C) 1.19 to 1.21 kg/l
Stabilizer ≤200 ppm phosphates
pH 1 to 4
Titration with KMnO4 49.8 to 50.6 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty