Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 35% (w/w) in water stabilized, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

44438-0050
TS44438-0050EA 108 USD InStock
TS44438-0050
Hydrogen peroxide 35% (w/w) in water stabilized, Technical Grade
Hydrogen peroxide


Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 108 °C (760 mmHg)
Melting Pt: –33 °C
Density: 1.135 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Additional info Stabilized
Titration with KMnO4 33 to 37 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty