Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide standard solution 1 N in water

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

12426-0010
TS12426-0010EA 40.74 USD InStock
TS12426-0010
Sodium hydroxide standard solution 1 N in water
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Density: 1.04 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.998 to 1.002 N (20°C)

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty