Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid ∼96% in water for analysis

Supplier: Thermo Scientific Chemicals
Danger

12464-0010 124640025 124645001 124645000 124640011 124640100 124640026
TS124645000EA 40.4 USD InStock
TS124645000 TS124645001 TS124640100 TS124640011 TS12464-0010 TS124640026 TS124640025
Sulfuric acid ∼96% in water for analysis
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Residue after ignition ≤0.0005 %
Titration with NaOH 95 to 97 %
Chloride (Cl) ≤0.5 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.5 ppm
Heavy metals (as Pb) ≤2 ppm
KMnO4-reducing substance ≤0.0008 % (as SO2)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty