Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Germanium(IV) chloride 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Germanium tetrachlorideTetrachlorogermane

AC19715-0250 AC19715-0050 19715-1000
TS19715-1000EA 793 USD InStock
TS19715-1000 TS19715-0250 TS19715-0050
Germanium(IV) chloride 99.99% (trace metals basis)
Germanium (IV) chloride


Formula: Cl₄Ge
MW: 214.42 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –49.5 °C
Density: 1.84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011029
CAS Number: 10038-98-9
EINECS: 233-116-7
UN: 2922
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Total trace metal impurities ≤0.01 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty