Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphoryl trichloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phosphorus oxychloridePhosphoryl chloridePhosphorus(V) oxychloridePhosphorus (V) oxide chloride

19129-0010
TS19129-0010EA 137.49 USD InStock
TS19129-0010
Phosphoryl trichloride 99%
Phosphoryl trichloride


Formula: POCl₃
MW: 153.33 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: 1.2 °C
Density: 1.645 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear liquid
Refractive index 1.4600 to 1.4620 (20°C, 589 nm)
Titration Argentometric ≥98.5 % (ex Cl)
Color scale ≤40 APHA
Phosphorus trichloride (PCl3) ≤0.3 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty