Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bronopol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromo-2-nitro-1,3-propanediol2-Bromo-2-nitropropane-1,3-diol

15882-5000 15882-1000 15882-0250
TS15882-0250EA 42.4 USD InStock
TS15882-0250 TS15882-1000 TS15882-5000
Bronopol 98%
Bronopol


Formula: HOCH₂BrC(NO₂)CH₂OH
MW: 199.99 g/mol
Melting Pt: >120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007390
CAS Number: 52-51-7
EINECS: 200-143-0
UN: 3241
ADR: 4.1,III
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point ≥120 °C
Titration Argentometric ≥97.5 % (Ex Br)
Water ≤0.5 %
GC ≥97.5 %
Iron (Fe) ≤0.001 %
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.005 %
Insoluble matter ≤0.005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty