Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia 7 N in methanol

Supplier: Thermo Scientific Chemicals
Danger

13371-0010 13371-0250 13371-0025
TS13371-0010EA 336.15 USD InStock
TS13371-0010 TS13371-0025 TS13371-0250
Ammonia 7 N in methanol
Ammonia


Formula: NH₃
MW: 17.03 g/mol
Density: 0.77 g/cm³
Flash Pt: 14 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011418
CAS Number: 7664-41-7
EINECS: 215-647-6
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Water ≤0.5 %
Strength 6.0 to 7.5 N
NMR (31P-NMR) Consistent with structure
Elemental Analysis 52.5 to 56.9 % C
Elemental Analysis 4.1 to 4.4 % N

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty