Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia 7N in methanol, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

42838-1000 42838-8000
TS42838-1000EA 78.78 USD InStock
TS42838-1000 TS42838-8000
Ammonia 7N in methanol, AcroSeal™
Ammonia


Formula: NH₃
MW: 17.03 g/mol
Boiling Pt: –33 °C (1013 hPa)
Melting Pt: –77 °C
Density: 0.77 g/cm³
Flash Pt: 14 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Strength 6.0 to 7.5 N
Refractive index 1.6110 to 1.6130 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty