Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia 0.5 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

38844-1000
TS38844-1000EA 105.71 USD InStock
TS38844-1000
Ammonia 0.5 M in THF, AcroSeal™
Ammonia


Formula: NH₃
MW: 17.03 g/mol
Density: 0.85 g/cm³
Flash Pt: –36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Solution
Water ≤2 % (K.F.)
Strength 0.40 to 0.60 M

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty