Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propiononitrile 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propionitrile

18089-5000 18089-1000 18089-0025
TS18089-1000EA 65.69 USD InStock
TS18089-1000 TS18089-0025 TS18089-5000
Propiononitrile 99%, Extra Pure
Propiononitrile


Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –93 °C
Density: 0.77 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001948
CAS Number: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3670 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty