Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroquinone 99.5%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,4-DihydroxybenzeneQuinol

21993-0051 21993-2500 21993-0010 21993-0500 21993-0050
TS21993-0500EA 28.57 USD InStock
TS21993-0500 TS21993-0010 TS21993-0051 TS21993-2500 TS21993-0050
Hydroquinone 99.5%
Hydroquinone


Formula: C₆H₄(OH)₂
MW: 110.11 g/mol
Boiling Pt: 285…287 °C (760 mmHg)
Melting Pt: 170…174 °C
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/01/78/2300178.jpg
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 171 to 174 °C
Water ≤0.6 % (K.F.)
Sulfated ash ≤0.05 %
HPLC ≥99.4 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty