Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Diaminonaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DAN2,3-Naphthalenediamine2,3-naphthylenediamine

15959-0010 15959-0250 15959-0050
TS15959-0050EA 867 USD InStock
TS15959-0050 TS15959-0250 TS15959-0010
2,3-Diaminonaphthalene 97%
2,3-Diaminonaphthalene


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Melting Pt: 193…199 °C
Density: 1.0968 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004116
CAS Number: 771-97-1
EINECS: 212-241-0
Order Now
Specification Test Results
Appearance Green-brown to brown Powder
Infrared spectrum Conforms
Melting point 193 to 199 °C
HPLC ≥96.0 %
Impurity ≤0.1 % (beta-Naphthylamine)
UV at 280 nm A: ≤0.035
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.03 %
Arsenic (As) ≤0.0001 %
Copper (Cu) ≤0.005 %
Iron (Fe) ≤0.005 %
Nickel (Ni) ≤0.005 %
Heavy metals (as Pb) ≤20 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty