Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobutan-1-ol 98%

Supplier: Thermo Scientific Chemicals
Danger

17635-0010 17635-0250 17635-0050
TS17635-0050EA 130 USD InStock
TS17635-0050 TS17635-0250 TS17635-0010
4-Aminobutan-1-ol 98%
4-Aminobutan-1-ol


Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 16…18 °C
Density: 0.96 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008230
CAS Number: 13325-10-5
EINECS: 236-364-4
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4630 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty