Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriethylsilane 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triethylchlorosilane

AC21594-1000 AC21594-0250 21594-5000
TS21594-0250EA 95.55 USD InStock
TS21594-0250 TS21594-5000 TS21594-1000
Chlorotriethylsilane 99%
Chlorotriethylsilane


Formula: C₆H₁₅ClSi
Boiling Pt: 142…144 °C (760 mmHg)
Density: 0.89 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty