Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexamethylditin 99%

Supplier: Thermo Scientific Chemicals
Danger

AC29585-0010 29585-0250 AC29585-0050
TS29585-0010EA 49 USD InStock
TS29585-0010 TS29585-0050 TS29585-0250
Hexamethylditin 99%
Hexamethylditin


Formula: C₆H₁₈Sn₂
MW: 327.63 g/mol
Boiling Pt: 182 °C (756 mmHg)
Melting Pt: 23…24 °C
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00008277
CAS Number: 661-69-8
EINECS: 211-549-2
UN: 3146
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear to slightly hazy colorless to white Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5410 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty