Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

N,N-Dimethylformamide, extra dry 99.8%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

32687-1000 32687-0010
TS32687-1000EA 82.18 USD InStock
TS32687-1000 TS32687-0010
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal™
N,N-Dimethylformamide


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.945 g/cm³
Flash Pt: 58 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.75 %
Residue after evaporation ≤0.001 %
Acidity (CH3COOH) ≤0.002 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty