Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid ∼70% in water, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

45177-0025
TS45177-0025EA 239 USD InStock
TS45177-0025
Perchloric acid ∼70% in water, Extra Pure
Perchloric acid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C (760 mmHg)
Melting Pt: –18 °C
Density: 1.66 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Appearance Clear solution
Color scale ≤10 APHA
Residue after ignition ≤0.003 %
Titration with NaOH 70 to 72 %
Chloride (Cl) ≤3 ppm
Sulfate (SO4) ≤5 ppm
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty