Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum(IV) chloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Platinum tetrachloride

AC19540-2500 AC19540-0010 19540-0050
TS19540-0010EA 163 USD InStock
TS19540-0010 TS19540-0050 TS19540-2500
Platinum(IV) chloride 99%
Platinum (IV) chloride


Formula: Cl₄Pt
MW: 336.89 g/mol
Melting Pt: 370 °C
Storage Temperature: Ambient
MDL Number: MFCD00011182
CAS Number: 13454-96-1
EINECS: 236-645-1
UN: 2923
ADR: 8,II
Order Now
Specification Test Results
Appearance Brown Powder and/or chunks
Trace metal analysis ≤500 ppm (Sum Ru+Rh+Ir+Pd)
Calcium (Ca) ≤250 ppm
Copper (Cu) ≤25 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤25 ppm
Magnesium (Mg) ≤250 ppm
Platinum (Pt) 57.0 to 58.5 %
Silicon (Si) ≤200 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty