Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid fuming (20 - 30% free SO₃), Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Oleum

41997-5000
TS41997-5000EA 244.56 USD InStock
TS41997-5000
Sulfuric acid fuming (20 - 30% free SO₃), Extra Pure
Sulfuric acid fuming


Formula: H₂O₇S₂ (H₂SO₄ + SO₃)
Melting Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00137792
CAS Number: 8014-95-7
UN: 1831
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Assay 20 to 30 % (free SO3)
Residue after ignition ≤0.01 %
Ammonium (NH4) ≤30 ppm
Arsenic (As) ≤2 ppm
Iron (Fe) ≤50 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty