Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tungstic (VI) acid 99+%

Supplier: Thermo Scientific Chemicals
Synonyms: Dihydrogen wolframateTungstic acid

20761-5000 AC20761-0050 AC20761-1000
TS20761-1000EA 77 USD InStock
TS20761-1000 TS20761-5000 TS20761-0050
Tungstic (VI) acid 99+%
Tungstic (VI) acid


Formula: H₂O₄W
MW: 249.85 g/mol
Density: 5.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2
Order Now
Specification Test Results
Appearance Yellow to yellow-green Powder
Assay ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty