Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilized, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

39662-1000
TS39662-1000EA 128.75 USD InStock
TS39662-1000
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilized, AcroSeal™
(±)-2-Methyltetrahydrofuran


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 78…80 °C (760 mmHg)
Melting Pt: –136 °C
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
UN: 2536
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.002 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm (BHT)
Peroxides ≤50 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty