Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,8-Bis(dimethylamino)naphthalene 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine

16986-0100 16986-0500
TS16986-0500EA 157 USD InStock
TS16986-0500 TS16986-0100
1,8-Bis(dimethylamino)naphthalene 99%
1,8-Bis(dimethylamino)naphthalene


Formula: C₁₄H₁₈N₂
MW: 214.31 g/mol
Boiling Pt: 144…145 °C (5hPa)
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003920
CAS Number: 20734-58-1
EINECS: 244-001-6
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance Light yellow to beige to pink or brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 46 to 50 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty