Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

(2-Hydroxylpropyl)-β-cyclodextrine 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Hydroxypropyl Betadex(2-Hydroxypropyl)-β-cyclodextrinHydroxypropyl-β-cyclodextrin

29756-5000 29756-0250 29756-1000
TS29756-1000EA 556.4 USD InStock
TS29756-1000 TS29756-5000 TS29756-0250
(2-Hydroxylpropyl)-β-cyclodextrine 97%
(2-Hydroxylpropyl)-β-cyclodextrine


Formula: C₆₃H₁₁₂O₄₂
Melting Pt: 278 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD16621721
CAS Number: 128446-35-5
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤7.5 % (K.F.)
HPLC ≥96.0 % (On dry substance)
HPLC ≤2.0 % Residual beta-cyclodextrin (On dry substance)
NMR 2 to 6 (degree of substitution)
NMR substituents are located mainly at O(2) of the anhydroglucopyranose units

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty