Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dimethylpyrrole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,4-Dimethyl-1H-pyrrole

31965-0250 31965-0010 31965-0050
TS31965-0010EA 43 USD InStock
TS31965-0010 TS31965-0050 TS31965-0250
2,4-Dimethylpyrrole 97%
2,4-Dimethylpyrrole


Formula: C₆H₉N
MW: 95.1424 g/mol
Boiling Pt: 165…167 °C
Density: 0.924 g/cm³
Flash Pt: >109 °C
Storage Temperature: Ambient
EINECS: 210-912-2
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4990 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty