Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

36828-0010 36828-0050
TS36828-0050EA 255 USD InStock
TS36828-0050 TS36828-0010
Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 98%
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate


Formula: C₆H₅Cl₂N₃O₂
MW: 222.03 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00010431
CAS Number: 1458-18-0
EINECS: 215-947-7
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Light brown to red-brown Powder
Infrared spectrum Conforms
Melting point 229 to 235 °C (Dec)
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty