Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Danger

20697-0050 20697-0010 20697-0250
TS20697-0250EA 1334 USD InStock
TS20697-0250 TS20697-0050 TS20697-0010
4-Methyl-1H-pyrazole 97%
4-Methyl-1H-pyrazole


Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 99…100 °C (6 mmHg)
Density: 0.99 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005245
CAS Number: 7554-65-6
EINECS: 231-445-0
UN: 1760
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty