Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Methoxyphenyl)magnesium bromide 1 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Anisylmagnesium bromide

AC37742-8000 37742-1000
TS37742-8000EA 274 USD InStock
TS37742-8000 TS37742-1000
(4-Methoxyphenyl)magnesium bromide 1 M in THF, AcroSeal™
(4-Methoxyphenyl)magnesium bromide


Formula: C₇H₇BrMgO
MW: 211.34 g/mol
Density: 1 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00013515
CAS Number: 13139-86-1
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.9 to 1.2 M

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty