Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

n-Heptane, extra dry 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

32691-0010
TS32691-0010EA 114.33 USD InStock
TS32691-0010
n-Heptane, extra dry 99+%, AcroSeal™
n-Heptane


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98 °C (760 mmHg)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.0005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty