Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Hexane ∼95% for HPLC

Danger
Synonyms: HexanenHexaneHexyl hydride

26836-0025
TS26836-0025EA 182 USD InStock
TS26836-0025
n-Hexane ∼95% for HPLC
n-Hexane


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.01 %
UV at 200 nm A: ≤0.70
UV at 210 nm A: ≤0.25
UV at 220 nm A: ≤0.07
UV at 230 nm A: ≤0.03
UV at 240 nm A: ≤0.01
UV at 250 nm A: ≤0.005
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Residue after evaporation ≤0.0002 %
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty