Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium carbonate 99+% ACS

Supplier: Thermo Scientific Chemicals
Synonyms: Marble

42351-5000 AC42351-1000 AC42351-0025
TS42351-0025EA 318 USD InStock
TS42351-0025 TS42351-1000 TS42351-5000
Calcium carbonate 99+% ACS
Calcium carbonate


Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Complexometric ≥99.0 % (on dried substance)
Chloride (Cl) ≤0.001 %
Fluoride (F) ≤0.0015 %
Sulfate (SO4) ≤0.01 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.01 %
Iron (Fe) ≤0.003 %
Magnesium (Mg) ≤0.02 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.1 %
Strontium (Sr) ≤0.1 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 % (in dilute HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty