Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% for analysis (max. 0.01 ppm Hg)

Supplier: Thermo Scientific Chemicals
Danger

45055-0025
TS45055-0025EA 83.1 USD InStock
TS45055-0025
Hydrochloric acid 37% for analysis (max. 0.01 ppm Hg)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 57 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Residue after ignition ≤0.0005 %
Titration with NaOH ≥37.0 %
Chloride (Cl) ≤0.5 ppm
Phosphate (PO4) ≤0.5 ppm
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Bromine (Br) ≤50 ppm
Aluminium (Al) ≤0.05 ppm
Ammonium (NH4) ≤1 ppm
Arsenic (As) ≤0.01 ppm
Barium (Ba) ≤0.02 ppm
Beryllium (Be) ≤0.02 ppm
Bismuth (Bi) ≤0.1 ppm
Cadmium (Cd) ≤0.01 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.01 ppm
Copper (Cu) ≤0.02 ppm
Germanium (Ge) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.02 ppm
Lithium (Li) ≤0.01 ppm
Magnesium (Mg) ≤0.01 ppm
Manganese (Mn) ≤0.1 ppm
Mercury (Hg) ≤0.01 ppm
Molybdenum (Mo) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Strontium (Sr) ≤0.01 ppm
Thallium (Tl) ≤0.05 ppm
Titanium (Ti) ≤0.1 ppm
Vanadium (V) ≤0.01 ppm
Zinc (Zn) ≤0.05 ppm
Zirconium (Zr) ≤0.1 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty