Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hyaluronic acid sodium salt 95%

Supplier: Thermo Scientific Chemicals
Synonyms: HA sodium saltSodium hyaluronate

25177-0250 25177-0010 25177-0050
TS25177-0010EA 102.99 USD InStock
TS25177-0010 TS25177-0250 TS25177-0050
Hyaluronic acid sodium salt 95%
Hyaluronic acid sodium salt


Formula: (C₁₄H₂₁NaNO₁₁)n
Storage Temperature: Refrigerator
MDL Number: MFCD00875848
CAS Number: 9067-32-7
Order Now
Specification Test Results
Appearance White to cream Powder
Infrared spectrum Conforms
Assay ≥94 % (On dry substance)
Loss on drying ≤10 % (500 mg, 105°C, 6 h)
Protein ≤0.2 %
Molecular weight 1.5 to 2.2 million Da
pH 6.0 to 8.0 (0.5 % in water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty