Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amberlite® IRA-402 (Cl) ion-exchange resin

Supplier: Thermo Scientific Chemicals

36994-0050
TS36994-0050EA 489 USD InStock
TS36994-0050
Amberlite® IRA-402 (Cl) ion-exchange resin
Amberlite® IRA-402 (Cl)


Storage Temperature: Ambient
MDL Number: MFCD00145564
CAS Number: 52439-77-7
Order Now
Specification Test Results
Appearance Light yellow Semitransparent beads
Particle size ≤1.0 % (< 300 micron)
Particle size ≤5.0 % (> 1180 micron)
Harmonic mean size 0.600 to 0.750 mm
Uniformity coefficient ≤1.60
Moisture holding capacity 49.0 to 60.0 % (Cl)
Total volume capacity ≥1.20 eq/l (Cl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty