Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 1X8 (Cl) 50-100 mesh ion-exchange resin

Supplier: Thermo Scientific Chemicals

20299-0025 20299-5000
TS20299-5000EA 416 USD InStock
TS20299-5000 TS20299-0025
Dowex® 1X8 (Cl) 50-100 mesh ion-exchange resin
Dowex® 1X8 (Cl)


Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 12627-85-9
Order Now
Specification Test Results
Appearance White to yellow to tan Beads (manufacturer - Does not impact use.)
Particle size ≤15 % (on 35 mesh, cumulative)
Particle size ≤15 % (through 80 mesh)
Wet volume capacity ≥1.2 meq/ml (Cl form)
Water retention capacity 43 to 48 % (Cl form)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty