Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 1X8 (Cl) 100-200 mesh ion-exchange resin

Supplier: Thermo Scientific Chemicals

20300-1000
TS20300-1000EA 126.58 USD InStock
TS20300-1000
Dowex® 1X8 (Cl) 100-200 mesh ion-exchange resin
Dowex® 1X8 (Cl)


Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 12627-85-9
Order Now
Specification Test Results
Appearance White to yellow Moist beads
Particle size ≤15 % (on 60 mesh, cumulative)
Particle size ≤15 % (through 170 mesh)
Wet volume capacity ≥1.2 meq/ml (Cl form)
Water retention capacity 39 to 45 % (Cl form)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty