Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 50WX8 (H) 100-200 mesh ion-exchange resin

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dowex 50 wx8

33534-1000 33534-0025
TS33534-0025EA 1528 USD InStock
TS33534-0025 TS33534-1000
Dowex® 50WX8 (H) 100-200 mesh ion-exchange resin
Dowex® 50WX8 (H)


Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 11119-67-8
Order Now
Specification Test Results
Appearance Beige to light brown Beads
Particle size ≤15 % (> 250 micron)
Particle size ≤15 % (< 88 micron)
Moisture holding capacity 50 to 58 % (H)
Total volume capacity ≥1.7 eq/l (H)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty