Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

61001-0040
TS61001-0040EA 445 USD InStock
TS61001-0040
Acetonitrile
Acetonitrile


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Refractive index 1.3405 to 1.3425 (25 °C, 589 nm)
Water ≤0.01 % (K.F.)
Additional info fluorescence background: passes test
GC ≥99.9 %
UV 1 cm cell vs water
UV at 190 nm A: ≤1
UV at 195 nm A: ≤0.15
UV at 200 nm A: ≤0.07
UV at 205 nm A: ≤0.05
UV at 210 nm A: ≤0.04
UV at 220 nm A: ≤0.02
UV at 254 nm A: ≤0.01
Color scale ≤10 APHA
Identification passes test
Specific gravity (25°C) 0.775 to 0.780
Residue after evaporation ≤1 ppm
Gradient analysis passes test
Titrable acid ≤0.008 meq/g
Titrable base ≤0.0006 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty