Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloromethane

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methane dichlorideMethylene chloride

61005-0040
TS61005-0040EA 235.41 USD InStock
TS61005-0040
Dichloromethane
Dichloromethane


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Refractive index 1.42 to 1.423 (25 °C, 589 nm)
Water ≤0.02 %
Additional info fluorescence background: passes test
GC ≥99.9 % Including preservative
UV 1 cm cell vs water
UV at 235 nm A: ≤1
UV at 240 nm A: ≤0.2
UV at 254 nm A: ≤0.014
Color scale ≤10 APHA
Identification passes test
Specific gravity (25 °C) 1.315 to 1.321
Residue after evaporation ≤2 ppm
Free halogens passes test
Titrable acid ≤0.0003 meq/g
Amylene 40 to 100 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty