Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.9% for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

61009-0040
TS61009-0040EA 204.89 USD InStock
TS61009-0040
Methanol ≥99.9% for HPLC
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Refractive index 1.3260 to 1.3300 (25 °C, 589 nm)
Water ≤0.1 %
Additional info formaldehyde < 0.001%
Additional info acetaldehyde < 0.001%
Additional info fluorescence background: passes test
Solubility (in water) Passes test
GC ≥99.9 %
UV 1 cm cell vs water
UV at 205 nm A: ≤1
UV at 220 nm A: ≤0.3
UV at 230 nm A: ≤0.15
UV at 254 nm A: ≤0.025
Color scale ≤5 APHA
Identification Passes test
Residue after evaporation ≤3 ppm
Gradient analysis Passes test
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
Acetone ≤0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty