Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 

Dichloromethane, extra dry, anhydrous ≥99.9%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methane dichlorideMethylene chloride

61093-1000
TS61093-1000EA 65.79 USD InStock
TS61093-1000
Dichloromethane, extra dry, anhydrous ≥99.9%, AcroSeal™
Dichloromethane


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Appearance Colorless Liquid
Assay ≥99.9 %
Water ≤50 ppm (K.F.)
Additional info inhibitor: 40 - 100 ppm amylene
Additional info inhibitor: 40 - 60 ppm cyclohexene

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty