Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome Black T solution

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonateEBTSolochrome Black TSodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

61222-5000
TS61222-5000EA 75 USD InStock
TS61222-5000
Eriochrome Black T solution
Eriochrome Black T


Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
UN: 1230
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear blue Solution
Identification passes test
Sensitivity passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty