Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calmagite, pure indicator grade

Supplier: Thermo Scientific Chemicals
Warning

10817-0050 10817-1000 10817-0250 10817-0100
TS10817-0250EA 144.03 USD InStock
TS10817-0250 TS10817-0050 TS10817-0100 TS10817-1000
Calmagite, pure indicator grade
Calmagite


Formula: C₁₇H₁₄N₂O₅S
MW: 358.37
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011656
CAS Number: 3147-14-6
EINECS: 221-563-0
Order Now
Specification Test Results
Appearance Black Crystalline powder
Infrared spectrum Conforms
Specific extinction E (1%/1cm) ≥300 (610 nm, in buffer)
Suitability (for Ca + Mg determination) Passes test
UV (2.7 g in 100 ml water) pH 3
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.05
UV at 230 nm A: ≤0.03
UV at 240 nm A: ≤0.02
UV at 250 nm A: ≤0.01
UV at 260 nm A: ≤0.01
Titration after Ion exchange ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty