Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenyliodonium chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

11733-0100 11733-0500
TS11733-0500EA 1133 USD InStock
TS11733-0500 TS11733-0100
Diphenyliodonium chloride 97%
Diphenyliodonium chloride


Formula: C₁₂H₁₀ClI
MW: 316.57 g/mol
Melting Pt: 227…228 °C (subl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011909
CAS Number: 1483-72-3
EINECS: 216-049-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
Chloride and Bromide ≤0.02 % (as Cl)
Iodide (I) passes test
Sulfate (SO4) ≤150 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.1 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty