Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Quinolinol ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 8-Hydroxyquinoline

12190-0250 12190-0025 12190-0010 12190-1000 12190-5000
TS12190-0250EA 57 USD InStock
TS12190-0250 TS12190-5000 TS12190-0010 TS12190-1000 TS12190-0025
8-Quinolinol ACS
8-Quinolinol


Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 267 °C (752 mmHg)
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
EINECS: 205-711-1
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/52/770552.jpg
Order Now
Specification Test Results
Appearance White to light yellow to light beige Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 72.5 to 74.0 °C
Residue after ignition ≤0.05 %
Suitability (for Mg determination) Passes test
Sulfate (SO4) ≤0.02 %
Insoluble matter ≤0.05 % (in alcohol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty