Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2-Nitrophenyl)methanol

12851-1000 12851-0050 12851-0250
TS12851-0250EA 64.01 USD InStock
TS12851-0250 TS12851-0050 TS12851-1000
2-Nitrobenzyl alcohol 97%
2-Nitrobenzyl alcohol


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 270 °C (760 mmHg)
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007186
CAS Number: 612-25-9
EINECS: 210-302-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty