Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propanal

13151-0100 13151-0250 13151-0025 13151-0010
TS13151-0100EA 668 USD InStock
TS13151-0100 TS13151-0010 TS13151-0025 TS13151-0250
Propionaldehyde 97%
Propionaldehyde


Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (760 mmHg)
Melting Pt: –81 °C
Density: 0.798 g/cm³
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 1208008
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥96.0 %
UV at 290 nm A: ≤0.04
UV at 280 nm A: ≤0.05
UV at 270 nm A: ≤0.10
UV at 260 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Titration with NaOH ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty