Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Chlorosuccinimide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Chloropyrrolidine-2,5-dione

14925-1000 14925-0025 14925-5000
TS14925-1000EA 31.43 USD InStock
TS14925-1000 TS14925-0025 TS14925-5000
N-Chlorosuccinimide 97%
N-Chlorosuccinimide


Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
Melting Pt: 144…150 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 144 to 150 °C
Titration Iodometric ≥96.0 % (on dry substance)
Loss on drying ≤4 % (2 g, 110 °C, 2 hrs)
Specific optical rotation +20.0° to +25.0° (20 °C, 589 nm) (c=1 in 8:2 AcOH/1M HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty