Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhodamine 6G, pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

19141-0500
TS19141-0500EA 46.12 USD InStock
TS19141-0500
Rhodamine 6G, pure
Rhodamine 6G


Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: 229.4 °C
MDL Number: MFCD00012665
CAS Number: 989-38-8
EINECS: 213-584-9
UN: 2811
Order Now
Specification Test Results
Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty